missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Mouse anti-Human, Clone: DMD/3243, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Dystrophin |
|---|---|
| Clone | DMD/3243 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Description
Dystrophin Monoclonal specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Dystrophin | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 1756 | |
| IgG1 κ | |
| Protein A or G purified |
| DMD/3243 | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
| A recombinant fragment (around aa 114-263) of human Dystrophin protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title