missing translation for 'onlineSavingsMsg'
Learn More

Dystrophin Mouse anti-Human, Clone: DMD/3243, Novus Biologicals™

Product Code. 18669757 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18669757 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18669757 Supplier Novus Biologicals Supplier No. NBP307641100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dystrophin Monoclonal specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dystrophin
Applications ELISA, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone DMD/3243
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142
Host Species Mouse
Immunogen A recombinant fragment (around aa 114-263) of human Dystrophin protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1756
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.