missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DYNLT1 Antibody (SR4309), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | DYNLT1 |
|---|---|
| Clone | SR4309 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041593
|
Novus Biologicals™
NBP3-27843-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041764
|
Novus Biologicals™
NBP3-27843-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DYNLT1 Monoclonal antibody specifically detects DYNLT1 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| DYNLT1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 6993 | |
| IgG | |
| Affinity purified |
| SR4309 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| CW-1, dynein light chain Tctex-type 1, dynein, light chain, Tctex-type 1, MGC111571, Protein CW-1, T-complex testis-specific protein 1 homolog, t-complex-associated-testis-expressed 1-like 1, TCTEL1tctex-1, TCTEX1, TCTEX-1 | |
| A synthesized peptide derived from human DYNLT1 (Uniprot #: P63172) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title