missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DYNLT1 Antibody (SR4309), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27843-100ul
This item is not returnable.
View return policy
Description
DYNLT1 Monoclonal antibody specifically detects DYNLT1 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| DYNLT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6993 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR4309 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| CW-1, dynein light chain Tctex-type 1, dynein, light chain, Tctex-type 1, MGC111571, Protein CW-1, T-complex testis-specific protein 1 homolog, t-complex-associated-testis-expressed 1-like 1, TCTEL1tctex-1, TCTEX1, TCTEX-1 | |
| A synthesized peptide derived from human DYNLT1 (Uniprot #: P63172) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu