missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC2H1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | DYNC2H1 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18108558
|
Novus Biologicals
NBP2-38437 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622596
|
Novus Biologicals
NBP2-38437-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DYNC2H1 Polyclonal specifically detects DYNC2H1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DYNC2H1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8NCM8 | |
| 79659 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LLSETLDLSELFHPDTFLNALRQETARAVGRSVDSLKFVASWKGRLQEAKLQIKISGLLLEGCSFDGNQLSENQLDSPSVSSVLPCFMGWIPQDA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ATD3, DHC1B, DHC2, DNCH2, DYH1B, dynein cytoplasmic heavy chain 2, dynein heavy chain 11, dynein, cytoplasmic 2, heavy chain 1, dynein, cytoplasmic, heavy polypeptide 2, FLJ11756, hdhc11, isotype 1B, KIAA1997 | |
| DYNC2H1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title