missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC2H1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38437-25ul
This item is not returnable.
View return policy
Description
DYNC2H1 Polyclonal specifically detects DYNC2H1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DYNC2H1 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q8NCM8 | |
| DYNC2H1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LLSETLDLSELFHPDTFLNALRQETARAVGRSVDSLKFVASWKGRLQEAKLQIKISGLLLEGCSFDGNQLSENQLDSPSVSSVLPCFMGWIPQDA | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ATD3, DHC1B, DHC2, DNCH2, DYH1B, dynein cytoplasmic heavy chain 2, dynein heavy chain 11, dynein, cytoplasmic 2, heavy chain 1, dynein, cytoplasmic, heavy polypeptide 2, FLJ11756, hdhc11, isotype 1B, KIAA1997 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79659 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction