Learn More
ABEBA™ Dynamic Flow White Occupational Sneakers O1
Occupational sneakers with white perforated microfiber, replaceable Active MediFit insole, slip resistant TPU outsole
Brand: ABEBA™ 7370/39
Additional Details : Weight : 0.71000kg
Description
- Breathable, lightweight, comfortable
- Comply to EN ISO 20347: 2012, O1 SRC
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- White perforated microfiber
- Breathable lining with silver ions
- Removable active Medifit insole
Specifications
White | |
39 | |
Women's | |
No |
Perforated Microfiber | |
Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340 | |
Sneakers | |
O1 SRC |