Learn More
Thermo Scientific™ DyLight™ 550 Maleimide
Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of maleimide-activated DyLight Dyes.
Brand: Thermo Scientific™ 62290
Code : PN
Additional Details : Weight : 0.00010kg
Description
Thermo Scientific™ Sulfhydryl-Reactive DyLight Dyes are maleimide-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as probes for cellular imaging and other fluorescence detection methods.
The Thermo Scientific™ DyLight™ Dyes have absorption spectra ranging from 350 to 770nm. These reagents fluoresce over a broad pH range, are more intense than Alexa Fluor™ or Cy™ Dyes in many applications and match the output wavelengths of common fluorescence instrumentation. Additionally, the water solubility of the DyLight Reagents allows a high fluor-to-protein ratio without precipitation during conjugation.The sulfhydryl-reactive dyes contain maleimide groups that react predominantly with free –SH groups at pH 6.5-7.5, forming a stable thioether bond. At pH 7, the maleimide group is ∼1000 times more reactive toward a free sulfhydryl than to an amine.1At pH values > 7.5, reactivity toward primary amines increases and hydrolysis of the maleimide group can occur.
Highlights:
- Specific — maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
- Efficient labeling methods — well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
- Optimized antibody labeling procedure —complete protocol for IgG reduction and labeling and calculating the labeling efficiency
Applications:
- Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
- Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies
Specifications
DyLight 550 Maleimide | |
Dye | |
Store desiccated at -20°C. Shipped at ambient temperature. | |
Maleimide | |
DyLight Fluors | |
576 |
1 mg | |
DMF (Dimethylformamide) | |
Thiol | |
DyLight™ 550 | |
DyLight™ | |
562 |
For Research Use Only. Not for use in diagnostic procedures.