missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ Orange Specialty Dyes

Product Code. p-4531918 Shop All Thermo Scientific Products
Click to view available options
Product Type:
DyLight 590-R2 NHS Ester
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 12351903

Brand: Thermo Scientific™ 46617

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Label amine-containing molecules for fluorescence imaging or flow cytometry with these rhodamine-based, orange-emitting (532 to 546 laser) NHS ester fluorescent dyes.

Thermo Scientific™ DyLight Orange-emitting Specialty Dyes are optimized for labeling molecules and other staining applications.

DyLight Orange-emitting Dyes are a family of labeling agents that provide bright fluorescent detection for imaging. Dyes can be selected based upon characteristic excitation and emission properties or relative hydrophilicity/hydrophobicity attributes. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetrating ability in vivo, while more hydrophilic dyes have less nonspecific binding potential. Each dye contains an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides or other biomolecules through amide bond formation.

Highlights:

  • NHS ester reactive group–allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation

  • Multiple solubility options –choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in a given application

Criteria to consider when choosing a DyLight Orange-emitting Specialty Dye:

  • Excitation and emission wavelengths –choose the best dye to match the excitation and emission capabilities of your instrument
  • Water solubility–choose a dye based on its relative hydrophilicity, which directly correlates to the number of negatively-charged sulfonates it has on its core structure. More hydrophilic dyes are best at maintaining water solubility of a labeled antibody and limiting the nonspecific binding of the conjugate. More hydrophobic dyes often are best at penetrating tissues and cell membranes in vivo, meaning that dyes with fewer sulfonates may work best for some applications.

Recommended for:

Imaging; Antibody labeling; Direct immunofluorescence staining; Flow cytometry; Fluorescence correlation spectroscopy; ELISA; Western blotting; Protein microarrays; Polymer labeling; Peptide labeling; Phalloidin labeling for actin staining; Staining in acidic media; Biofilm microorganism staining

TRUSTED_SUSTAINABILITY

Specifications

Formulation NHS ester activated Rhodamine derivative with 2 sulfonate groups
Quantity 1 mg
Product Type DyLight 590-R2 NHS Ester
Sufficient For 1 use to label a total of 7mg of IgG using typical conditions
Product Line DyLight
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.