missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 521-LS NHS Ester

Product Code. 12381943 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 12381943

Brand: Thermo Scientific™ 82495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Label antibodies or other proteins for FRET and multiplex fluorescence assays using fluors with large Stokes shifts (488nm excitation; yellow to far-red emission).

Thermo Scientific™ DyLight Long Stokes Shift Specialty Dyes offer a broad spectrum of fluorescence emission characteristics from green to far red with a single excitation peak that matches the 488 laser line and extraordinarily large Stokes shifts that inhibit quenching.

DyLight Long Stokes Shift Specialty Dyes are a family of labeling agents that provide bright fluorescent detection for multiplexed imaging. The dyes share a common excitation range, while their emission properties extend from the green to the far red wavelengths. The dyes are based on a core coumarin structure that is modified to emit at different wavelengths. Each dye contains an amine-reactive NHS ester for rapid modification of antibodies, proteins, peptides or other biomolecules through amide bond formation. DyLight Long Stokes Shift Dyes can be used individually or combined with DyLight 488 for multiplexed analysis. The excitation and emission peaks are sufficiently separated so as to minimize dye-dye quenching in assays.

Highlights:

  • Single excitation wavelength–DyLight Long Stokes Shift Dyes all can be excited together at 488nm using an Argon laser

  • Image multiple targets simultaneously–the four Long Stokes Shift Dyes can be used with DyLight 488 for multiplex fluorescence applications

  • NHS ester reactive group–allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation

  • Multiple solubility options –choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in an application

  • Large Stokes shift inhibits quenching–minimal overlap between excitation and emission peaks inhibits dye-dye quenching effects

  • Use in FRET-based assays– the extremely broad separation between excitation and emission peaks allow for use of the Long Stokes Shift Dyes in energy transfer applications where the acceptor dye is spectrally far removed from the excitation wavelength

Criteria to consider when choosing a DyLight Long Stokes Shift Specialty Dye:

  • Excitation and emission wavelengths–all dyes share a common excitation wavelength range with options for emission from the green to the far red wavelengths

  • Water solubility–DyLight Long Stokes Shift Dyes can be initially solubilized in methanol, ethanol, DMF or DMSO with transfer into an aqueous buffer medium for labeling biomolecules

Recommended for:

Quenchers and FRET pairs; Conjugation to silica particles; STED microscopy; Multicolor microscopy; 4Pi microscopy; Dual-color 3D nanoscopy; Dynamic light scattering; Graded stokes shift; Duplex enhanced RT-PCR; High-resolution optical microscopy

TRUSTED_SUSTAINABILITY

Specifications

Description DyLight 521-LS NHS Ester
Quantity 1 mg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store at -20°C. Shipped at ambient temperature.
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye DyLight™ Fluor (Specialty)
Label Type DyLight Fluors
Product Line DyLight
Emission 666
Excitation 526
Show More Show Less
compliance-icons
Product Identifier
  • DyLight 521-LS NHS Ester
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : DyLight Reactive Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.