missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 650-Phosphine

Product Code. 11387431 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387431 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11387431 Supplier Thermo Scientific™ Supplier No. 88911

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Label and detect azide-tagged molecules for metabolic labeling (Staudinger ligation) for fluorescence assays with phosphine-activated fluorescent dyes.

Thermo Scientific™ DyLight Fluor Phosphine Reagents are phosphine-activated fluorescent dyes for specific labeling and detection of azide-tagged molecules, which enables use of fluorescence imaging in metabolic labeling strategies.

Azide groups, which are the reactive targets of the phosphine-activated fluors, can be introduced into proteins or other cellular targets via azide reagents or through in vivo labeling with azide-derivatives of naturally occurring metabolic building blocks. Because neither phosphines nor azides are present in biological systems, they comprise chemoselective (mutually specific) ligation pairs for labeling and conjugation. Thus, when used in combination with azide labeling strategies, Phosphine-Activated DyLight Fluors enable directed fluorescent labeling to facilitate detection of protein interactions and post-translational modifications using fluorescence imaging technologies.

Highlights:

  • Soluble easily dissolves in water-miscible solvents (e.g., DMSO) for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples

  • Compatible reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents, and does not interfere with fluorescence applications

  • Chemoselective the phosphine reactive group is specific in biological samples for azide-tagged molecules, ensuring that fluorescent labeling is specific

  • High-performance fluorescence DyLight 488, DyLight 550 and DyLight 650 are intense, highly stable fluorophores for green, orange and red fluorescent detection

TRUSTED_SUSTAINABILITY

Specifications

Formulation Phosphine-activated proprietary far-red fluorescent dye
Product Type 650-Phosphine
Quantity 1 mg
Content And Storage Store desiccated at -20°C. Shipped at ambient temperature.
Product Line DyLight
Chemical Reactivity Azide
Reactive Moiety Phosphine
Solubility DMF (Dimethylformamide)
Label or Dye DyLight 650
Label Type Staudinger Chemistry Label, DyLight Fluors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.