Learn More
Description
Thermo Scientific™ DyLight Fluor Phosphine Reagents are phosphine-activated fluorescent dyes for specific labeling and detection of azide-tagged molecules, which enables use of fluorescence imaging in metabolic labeling strategies.
Highlights:
Soluble easily dissolves in water-miscible solvents (e.g., DMSO) for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples
Compatible reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents, and does not interfere with fluorescence applications
Chemoselective the phosphine reactive group is specific in biological samples for azide-tagged molecules, ensuring that fluorescent labeling is specific
High-performance fluorescence DyLight 488, DyLight 550 and DyLight 650 are intense, highly stable fluorophores for green, orange and red fluorescent detection
Specifications
Specifications
| Formulation | Phosphine-activated proprietary orange fluorescent dye |
| Product Type | 550-Phosphine |
| Quantity | 1 mg |
| Content And Storage | Store desiccated at -20°C. Shipped at ambient temperature. |
| Product Line | DyLight |
| Chemical Reactivity | Azide |
| Reactive Moiety | Phosphine |
| Solubility | DMF (Dimethylformamide) |
| Label or Dye | DyLight 550 |
| Label Type | Staudinger Chemistry Label, DyLight Fluors |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.