Learn More
Description
Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.
Highlights:
Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate
Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)
Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | Dye |
| Solubility | DMF (Dimethylformamide) |
| Content And Storage | Store desiccated at -20°C. |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Label or Dye | DyLight™ 650 |
| Label Type | DyLight Fluors |
| Product Line | DyLight |
| Emission | 672 |
| Show More |
Safety and Handling
- DyLight Reactive Dyes
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: DyLight Reactive Dyes
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.