Learn More
Thermo Scientific™ DyLight™ 633 Microscale Antibody Labeling Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ DyLight Fluor Antibody Labeling Kits provide the essential reagents and accessories to easily label and purify antibodies and include regular-scale and microscale kits for each specific DyLight Fluorescent Dye.
Highlights:
- Fast - label and purify protein in approximately one hour
- Amine-reactive dyes - label virtually any protein
- Efficient non-reacted dye removal
- Minimal sample dilution
- Spin column format eliminates the need for column preparation, fraction screening and waiting for protein to emerge from column
Item-Specific Features:
- Standard kits contain sufficient reagents to label and purify 3 × 1mg of IgG or similar quantities of other proteins
- Microscale kits contain sufficient reagents to label and purify 5 × 0.1mg of IgG or similar quantities of other proteins
Includes:
Antibody Labeling Kits
DyLight NHS Ester, 3 vials; Reaction buffer, 1mL; Spin columns, 6 each; Microcentrifuge collection tubes, 12 each; Purification resin, 5mL
Microscale Kits
DyLight NHS Ester, 5 vials; Reaction buffer, 1mL; Spin columns, 5 each; Microcentrifuge collection tubes, 10 each; Purification resin, 5mL
Specifications
Specifications
| Description | DyLight 633 Antibody Labeling Microscale Kit |
| Product Type | Antibody Labeling Kit |
| Quantity | 5 Reactions |
| Sufficient For | Labeling and purifying 5 x 100 μg (1 mg/mL) of IgG or other proteins |
| Content And Storage | Sufficient For: Labeling and purifying 5 x 100 μg (1 mg/mL) of IgG or other proteins Store the DyLight NHS Ester at -20°C. Store all other kit components at 4°C. |
| Excitation/Emission | 638/658 |
| Labeling Scale | 100 μg |
| Product Line | DyLight™ |
| Label or Dye | DyLight 633 |
| Labeling Target | Antibodies, Proteins |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.