missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 594 NHS Ester

Product Code. 11879370 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
5 x 65 μg
Unit Size:
1mg
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11879370 5 x 65 μg Pack of 5
10127833 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11879370 Supplier Thermo Scientific™ Supplier No. 46413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of NHS ester-activated DyLight Dyes.

Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.

The DyLight Dyes exhibit higher fluorescence intensity and photostability than Alexa™, CyDye™ and LI-COR™ Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9). Additionally, the water solubility of the DyLight Dyes allows a high dye-to-protein ratio without precipitation during conjugation.

Highlights:

  • Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate

  • Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)

  • Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 65 μg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C.
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye DyLight™ 594
Label Type DyLight Fluors
Product Line DyLight
Emission 618
Excitation 593
Labeling Target Proteins
Show More Show Less
compliance-icons
Product Identifier
  • DyLight 594 NHS Ester 5 x 65 ug
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: DyLight Reactive Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.