missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 350 NHS Ester

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of NHS ester-activated DyLight Dyes.

Brand:  Thermo Scientific™ 46426

Code : PN

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 10482074

  • £286.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.

The DyLight Dyes exhibit higher fluorescence intensity and photostability than Alexa™, CyDye™ and LI-COR™ Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9). Additionally, the water solubility of the DyLight Dyes allows a high dye-to-protein ratio without precipitation during conjugation.

Highlights:

  • Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate

  • Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)

  • Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation

Specifications

Specifications

DyLight 350 NHS Ester
Dye
Store desiccated at -20°C, protected from light.
Active Ester, Succinimidyl Ester, NHS Ester
DyLight Fluors
432
Proteins
1 mg
DMF (Dimethylformamide)
Amine
DyLight™ 350
DyLight™
353
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.