missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27327-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1 and ERK2, is predomitly expressed in hematopoietic tissues, and is localized in the nucleus.Specifications
| DUSP2 | |
| Polyclonal | |
| Unconjugated | |
| DUSP2 | |
| dual specificity phosphatase 2, DUSP2, PAC 1, PAC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13537, 1844 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q05922, Q05923 | |
| DUSP2 | |
| DUSP2 Fusion Protein Ag25380 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction