Learn More
DWK Life Sciences DURAN™ Vacuum Receiver Adapter, straight, with 2 standard ground joints, and screw-on plastic hose connection
Shop All DWK Life Sciences ProductsDescription
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask. If vacuum distillation is performed, the vacuum pump is ideally connected to the receiver adapter.
- Tight closure due to standard ground joint
- Straight
- With two standard ground joint and screw-on plastic hose connection
- Fired-on, highly durable white print
Specifications
Specifications
| Description | Autoclavable |
| For Use With (Application) | Distillation |
| Fits Tubing ID (Metric) | 8.6 mm |
| Material | Borosilicate Glass (3.3), PBT, Polypropylene, Silicone |
| Quantity | 1 Ea. |
| Thread Size | NS 14/23 |