Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
Product Code. 15395817 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
GL 14, with Cone NS 14/23
GL 14, with Cone NS 19/26
GL 14, with Cone NS 24/29
GL 14, with Cone NS 29/32
GL 18, with Cone NS 14/23
GL 18, with Cone NS 19/26
GL 18, with Cone NS 24/29
GL 18, with Cone NS 29/32
GL 25, with Cone NS 19/26
GL 25, with Cone NS 24/29
GL 25, with Cone NS 29/32
GL 32, with Cone NS 24/29
GL 32, with Cone NS 29/32
Unit Size:
Each
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No unitSize
15395817 GL 18, with Cone NS 14/23 Pack of 10
13427467 GL 14, with Cone NS 14/23 Each
13595370 GL 14, with Cone NS 19/26 Pack of 10
15222009 GL 14, with Cone NS 29/32 Pack of 10
15305827 GL 18, with Cone NS 29/32 Pack of 10
15282009 GL 25, with Cone NS 29/32 Pack of 10
15292009 GL 32, with Cone NS 29/32 Pack of 10
15242009 GL 18, with Cone NS 19/26 Pack of 10
15272009 GL 25, with Cone NS 24/29 Pack of 10
11639978 GL 32, with Cone NS 24/29 Pack of 10
12799696 GL 18, with Cone NS 24/29 Pack of 10
15656935 GL 25, with Cone NS 19/26 Pack of 10
12789696 GL 14, with Cone NS 24/29 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15395817 Supplier DWK Life Sciences Supplier No. 248416109
Only null left
Add to basket
Add to basket
The threaded tube with thread and standard ground joint allows the bonding of various glass components.
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
  • Tight closure due to standard ground joint
  • With DIN thread
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Item Description Tube with screw threads
Type Non-DIN Size
Standard Taper Joint No GL 18, with Cone NS 14/23