missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Triple-Neck Round Bottom Flask, with standard ground joint, parallel side necks

Multi-neck pellets are used for distillation, fraction and reactions of all kinds.

Brand:  DWK Life Sciences 241855509

Code : NEW

Additional Details : Weight : 0.35700kg

 View more versions of this product

Product Code. 10468572

  • £91.60 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.

  • Tight closure due to standard ground joint
  • Parallel side necks
  • Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
  • The geometry permits very uniform heating
  • Fired-on, highly durable white print
Specifications

Specifications

131 mm
Borosilicate Glass (3.3)
1/Pk.
Yes
1000 mL
Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volume sizes 250 mL (NS 24/29 and NS 29/32), 500 mL (NS 24/29), 500 mL (NS 29/32), 1000 mL (NS 29/32) and 2000 mL (NS 29/32) are not according to DIN ISO
Parallel side necks NS 29/32
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers