Learn More
DWK Life Sciences DURAN™ Triple-Neck Round Bottom Flask, with standard ground joint, parallel side necks
Multi-neck pellets are used for distillation, fraction and reactions of all kinds.
£64.65 - £128.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volume sizes 250 mL (NS 24/29 and NS 29/32), 500 mL (NS 24/29), 500 mL (NS 29/32), 1000 mL (NS 29/32) and 2000 mL (NS 29/32) are not according to DIN ISO |
No. per Pack | 1/Pk. |
Autoclavable | Yes |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
10171781
|
DWK Life Sciences
241854401 |
500 mL | Parallel side necks NS 19/26 |
£97.65
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
10478572
|
DWK Life Sciences
241853705 |
250 mL | Parallel side necks NS 14/23 |
£80.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10577281
|
DWK Life Sciences
241854504 |
500 mL | Parallel side necks NS 29/32 |
£102.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
10468572
|
DWK Life Sciences
241855509 |
1000 mL | Parallel side necks NS 29/32 |
£91.60
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10537471
|
DWK Life Sciences
241855603 |
1000 mL | Parallel side necks NS 14/23 |
£75.30
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10567281
|
DWK Life Sciences
241856505 |
2000 mL | Parallel side necks NS 29/32 |
£103.00
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10408962
|
DWK Life Sciences
241854607 |
500 mL | Parallel side necks NS 14/23 |
£64.65
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
10557281
|
DWK Life Sciences
241856308 |
2000 mL | Parallel side necks NS 14/23 |
£128.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10613792
|
DWK Life Sciences
241853602 |
250 mL | Parallel side necks NS 19/26 |
£72.65
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
Description
For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.
- Tight closure due to standard ground joint
- Parallel side necks
- Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
- The geometry permits very uniform heating
- Fired-on, highly durable white print
Specifications
Borosilicate Glass (3.3) | |
Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volume sizes 250 mL (NS 24/29 and NS 29/32), 500 mL (NS 24/29), 500 mL (NS 29/32), 1000 mL (NS 29/32) and 2000 mL (NS 29/32) are not according to DIN ISO | |
1/Pk. | |
Yes |