missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ TILT Thread Adaptor, GL 45 / GL 56, White PTFE

Product Code. 15563537 Shop All DWK Life Sciences Products
Change view
Click to view available options
White PTFE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15563537 White PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15563537 Supplier DWK Life Sciences Supplier No. 291195601
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

GL 45/GL 56 adaptor compatible with bottle top filter units.

The re-usable adaptor (GL 45 external / GL 56 internal) allows the use of the DURAN™ TILT bottle with 45 mm filtration units for the filter sterilisation of cell culture media. Manufactured from inert PTFE; can be autoclaved and at 300°C.

  • The reusable adaptor (GL 45 external/GL 56 internal) allows the use of the DURAN™ TILT bottle with 45 mm filter units for the filter sterilisation of cell culture media
  • Allows any 45 mm filter units to be used with DURAN™ TILT bottle
  • Manufactured from inert PTFE
TRUSTED_SUSTAINABILITY

Specifications

Color White
Diameter (Metric) Base 65 mm
Material PTFE
For Use With (Application) Allows any 45 mm filter units to be used with DURAN™ TILT bottle; Vacuum filter sterilization
No. per Pack 1/Pk.
Thread Size GL 45 / GL 56
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.