Learn More
DWK Life Sciences DURAN™ Straight Burette Valve, GU
Product Code. p-7189382 Shop All DWK Life Sciences Products
Click to view available options
For Use With (Equipment):
For Burette 50 mL
Unit Size:
Pack of 5
Product Code. 11600049
Brand: DWK Life Sciences 286059208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Through burette valve as an economical alternative.
Valves GU have an unscrewing safety device which prevents the valve pin from being unscrewed absolutely. The adjustable valve pin increases the service life of the valve tap. GU Valves are suitable for lower leakage rate requirements, e.g. for flow control of liquids or gases.
  • Through burette valve valve as low-cost alternative
  • Media flowing through the valve only come into contact with DURAN™ glass and PTFE.
  • Grease-free working
  • Handy design of the valve allows easy disassembly
  • The unscrewing protection prevents absolute unscrewing of the valve pin.
  • Valve pin adjustable to increase the service life of the valve tap
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3), PTFE
Autoclavable Autoclavable
No. per Pack 10/Pk.
For Use With (Application) GU straight through burette valve valves are suitable for lower leakage rate requirements, e.g. for flow control of liquids; Ideal for dosing and controlling liquids
For Use With (Equipment) For Burette 50 mL