Learn More
DWK Life Sciences DURAN™ Straight Burette Valve, GU
Shop All DWK Life Sciences Products
Click to view available options
For Use With (Equipment):
For Burette 50 mL
Unit Size:
Pack of 5
Description
Valves GU have an unscrewing safety device which prevents the valve pin from being unscrewed absolutely. The adjustable valve pin increases the service life of the valve tap. GU Valves are suitable for lower leakage rate requirements, e.g. for flow control of liquids or gases.
- Through burette valve valve as low-cost alternative
- Media flowing through the valve only come into contact with DURAN™ glass and PTFE.
- Grease-free working
- Handy design of the valve allows easy disassembly
- The unscrewing protection prevents absolute unscrewing of the valve pin.
- Valve pin adjustable to increase the service life of the valve tap
- Glass articles for further processing
Specifications
Specifications
| Material | Borosilicate Glass (3.3), PTFE |
| Autoclavable | Autoclavable |
| No. per Pack | 10/Pk. |
| For Use With (Application) | GU straight through burette valve valves are suitable for lower leakage rate requirements, e.g. for flow control of liquids; Ideal for dosing and controlling liquids |
| For Use With (Equipment) | For Burette 50 mL |