missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Silicone Septum Seal, Peroxide Cured Silicone, Septum for Piercing, Use with Open Topped Screw Cap
Shop All DWK Life Sciences ProductsDescription
- Compatible with open topped GL screw caps with central aperture
- Compatible with DURAN GL bottles
- Peroxide catalysed silicone construction
- Heat resistant to 130°C (steam) and 200°C (dry heat)
- Designed for injection or removal of media via piercing
Specifications
Specifications
| Color | Translucent |
| For Use With (Application) | Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments |
| Material | Silicone |
| Quantity | 100/Pk. |
| Sterility | Non-sterile |
| Thickness (Metric) | 2 mm |
| Thread Size (Metric) | GL 18 |
| Diameter (Metric) | 16 mm |
| Autoclavable | Autoclavable |
| Shape | Round |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?