missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Silicone Septum Seal, Peroxide Cured Silicone, Septum for Piercing, Use with Open Topped Screw Cap

Product Code. 11717198 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
11717198 GL 25 Pack of 100
13177374 GL 45 Pack of 100
13187164 GL 32 Pack of 100
11707198 GL14 Pack of 100
11787198 GL 18 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11717198 Supplier DWK Life Sciences Supplier No. 292460906
  • ONLINE EXCLUSIVE PRICE
    £158.62
    £126.09 / Pack of 100
    Save £32.53
    20% Off
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DURAN™ Silicone Septum Seal, Peroxide Cured Silicone, Septum for Piercing, Use with Open Topped Screw Cap is suitable for PBT open topped screw caps with central aperture. Peroxide catalysed silicone offers heat resistance to 130°C (steam) and 200°C (dry heat).

  • Compatible with open topped GL screw caps with central aperture
  • Compatible with DURAN GL bottles
  • Peroxide catalysed silicone construction
  • Heat resistant to 130°C (steam) and 200°C (dry heat)
  • Designed for injection or removal of media via piercing

Specifications

Color White
For Use With (Application) Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments
Material Silicone
Quantity 100/Pk.
Sterility Non-sterile
Thickness (Metric) 2 mm
Thread Size (Metric) GL 25
Diameter (Metric) 22 mm
Autoclavable Autoclavable
Shape Round
Product Type Septa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.