Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, SVL
To attach.
Brand: DWK Life Sciences 286266405
Additional Details : Weight : 25.00000kg
Description
The DURAN™ glass thread tube with SVL thread is a finer thread than the GL thread, which is also used for attaching to glass apparatus. The nominal size refers to the core diameter at the thread.
- Glass thread tube with SVL thread is a finer thread compared to GL thread
- The nominal size refers to the core diameter at the thread.
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing
Specifications
50/Pk. | |
32 mm | |
2 mm |
For attachment to glass apparatuses; Installation in glass apparatus and devices | |
SVL 30 |