Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, SVL
To attach.
£78.30 - £222.00
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
15804638
|
DWK Life Sciences
286266208 |
SVL 22 |
£168.00
Pack of 50 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
15894628
|
DWK Life Sciences
286265006 |
SVL 15 |
£78.30
Pack of 50 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
15814638
|
DWK Life Sciences
286266405 |
SVL 30 |
£222.00
Pack of 50 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
15844548
|
DWK Life Sciences
286064408 |
SVL 42 |
£121.00
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
The DURAN™ glass thread tube with SVL thread is a finer thread than the GL thread, which is also used for attaching to glass apparatus. The nominal size refers to the core diameter at the thread.
- Glass thread tube with SVL thread is a finer thread compared to GL thread
- The nominal size refers to the core diameter at the thread.
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing