Learn More
DWK Life Sciences DURAN™ Screw Thread Coupling, from PBT
Shop All DWK Life Sciences Products
Click to view available options
Thread Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Description
The screw thread coupling consists of two threaded caps, which are connected to each other via a flexible hose piece. This component enables a flexible connection of two glass components.
- For flexible connection of two glass screw-thread connections with integral silicone seal (VQM)
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | PBT |
| For Use With (Application) | Connection of glass threads |
| Packaging Quantity | 1 Ea. |
| Thread Size | GL 18 |