missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Round Bottom Flask, wide neck, with beaded rim, non-DIN EN ISO size

Product Code. 15241789 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
20,000 mL
2000 mL
3000 mL
50 mL
5000 mL
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15241789 2000 mL Pack of 10
11726093 50 mL Pack of 10
10709491 3000 mL Each
10241761 5000 mL Each
10261901 10,000 mL Each
10455041 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15241789 Supplier DWK Life Sciences Supplier No. 217416409
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Due to their spherical shape, round bottom flasks are used for heating liquids, among other things.

Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.

  • The wide neck enables easy filling and cleaning
  • The geometry permits very uniform heating
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Base 166 mm
Capacity (Metric) 2000 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Suitable for heating mantles; Heating; Mixing several components; Sizes 50, 1000, 2000, 3000, 5000, 6000, 10000, 12000 and 20000 mL non-DIN ISO size
No. per Pack 10/Pk.
Product Type Non DIN EN ISO size
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.