Learn More
DWK Life Sciences DURAN™ Reduction Adapter, socket/cone
Product Code. 15884818 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
Socket NS 19/26, Cone NS 14/23
Socket NS 24/29, Cone NS 14/23
Socket NS 24/29, Cone NS 19/26
Socket NS 29/32, Cone NS 14/23
Socket NS 29/32, Cone NS 19/26
Socket NS 29/32, Cone NS 24/29
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Size unitSize
15884818 Socket NS 29/32, Cone NS 14/23 Pack of 10
10223881 Socket NS 29/32, Cone NS 24/29 Each
10305101 Socket NS 29/32, Cone NS 19/26 Each
15981601 Socket NS 24/29, Cone NS 19/26 Pack of 10
15961601 Socket NS 19/26, Cone NS 14/23 Pack of 10
15844818 Socket NS 24/29, Cone NS 14/23 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15884818 Supplier DWK Life Sciences Supplier No. 286548004
Only null left
Add to basket
Add to basket
Reduction from sleeve to core.
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
  • Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Item Description Connection of two components with different standard ground joint sizes
Joint Size Socket NS 29/32, Cone NS 14/23