missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ DURAN™ protect+ Graduated Clear Laboratory Bottle with Wide Mouth
GREENER_CHOICE

Product Code. 17906762 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
3.5 L
5 L
Unit Size:
Each
Pack of 10
Pack of 6
Product Code. Capacity (Metric) unitSize
17906762 2 L Pack of 6
17966752 250 mL Pack of 10
17996752 500 mL Pack of 10
17926762 1 L Pack of 10
17936772 3.5 L Each
17966762 5 L Each
6 options
This item is not returnable. View return policy

Product Code. 17906762

Brand: DWK Life Sciences™ 218656382

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Multi-functional glass wide-mouth laboratory bottles with protective safety coating.

The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.

A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.

  • Manufactured from Polyamide-coated DURAN™ borosilicate glass 3.3
  • DURAN™ is a neutral glass, classified as a Glass Type I according to USP, EP (3.2.1) and JP, and meets ASTM E438, Type I, Class A requirements
  • Reduce the hazards of broken glass and liquid splashes in the event of breakage
  • The external polymer coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL and less)
  • Polyamide 12 safety coating has a high chemical resistance
  • Protective coating provides an enhanced safer grip
  • Coating is Cleanroom certified Fraunhofer IPA TESTED DEVICE™
  • Certified FDA and EU food contact Safe coating material
  • Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
  • Supplied with push-on dust cover
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 L
Capacity (English) 67.6 oz.
Color Clear
For Use With (Application) Preparation of media and reagents, safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs), sample or harvest collection container, culturing vessel, storage of media and reagents, transport and shipping, Liquid handling reservoir
Material Borosilicate Glass (3.3)
Quantity 6 Bottles
Sterility Non-sterile
Weight (Metric) 1.145 kg
Thread Size (Metric) GLS 80
Certifications/Compliance CoC
Closure Included No
Graduated Yes
Mouth Wide
Shape Round
Height (Metric) 248 mm
Product Type Laboratory Bottle
Closure Size 80
Diameter (Metric) 136 mm
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.