missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ DURAN™ protect+ Graduated Amber Laboratory Bottle with Wide Mouth
GREENER_CHOICE

Product Code. 17986752 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
3.5 L
5 L
Unit Size:
Each
Pack of 10
Pack of 6
This item is not returnable. View return policy

Product Code. 17986752

Brand: DWK Life Sciences™ 218664485

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Multi-functional glass wide-mouth laboratory bottles with protective safety coating.

The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.

A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.

  • Manufactured from ambered borosilicate glass 3.3 and polyamide
  • Compatible with matching cap system for particularly tight sealing and simple, clean pouring out
  • protect+ safety coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
  • UV protection USP (Spectral Transmission) and EP (3.2.1) compliant
  • Less than 10% spectral transmission between 290 nm and 450 nm
  • The only external application of the amber colour and safety coating ensures that internal surfaces have standard DURAN™ glass properties
  • Supplied as safety coated glass bottle only (screw caps and pouring rings must be ordered separately)
  • Easy-to-read scale with graduation marks to estimate the content
  • Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
  • Coating process does not use any volatile organic compounds (VOCs) or isocyanates
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Amber
For Use With (Application) Preparation of light-sensitive media and reagents, safe storage and shipping of bulk, light-sensitive, high-potency active pharmaceutical ingredients (HPAPIs) powders, sample or harvest collection container, sulturing vessel, storage of light-sensitive media and reagents, transport and shipping
Material Borosilicate Glass (3.3)
Quantity 10 Bottles
Sterility Non-sterile
Weight (Metric) 0.533 kg
Thread Size (Metric) GLS 80
Certifications/Compliance CoC
Closure Included No
Graduated Yes
Mouth Wide
Shape Round
Height (Metric) 148 mm
Product Type Laboratory Bottle
Closure Size 80
Diameter (Metric) 101 mm
Autoclavable Not Autoclavable
Show More Show Less
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt