Learn More
DWK Life Sciences DURAN™ Premium Screw Cap, Colourless PFA, with PTFE-coated (Platinum cured) silicone cap liner
Shop All DWK Life Sciences ProductsDescription
Thanks to its highly resistant materials, the premium cap is optimally suited to utilize in processes which are both chemically and thermally demanding. Due to its high temperature resistance, it is ideal for demanding processes, such as hot-air / dry-heat sterilisation and depyrogenation at 200°C. The materials used meet the requirements of the FDA/USP/EP and JP. Consequently the premium cap is particularly suitable for use in the pharmaceutical industry. Due to the PTFE-coated silicone cap liner, the premium cap provides a reliable and tight seal. Therefore it is ideal for the long term storage and transport of sensitive products. The matching PFA pouring ring permits drip-free pouring is available separately. Replacement PTFE faced silicone cap liners are available (platinum catalysed silicone)
- High purity polymer without any colourants
- USP Class VI Pharmaceutical grade materials
- Fluoropolymer cap offers high temperature resistance
- Fluoropolymer cap offers excellent chemical resistance
- High tightness due to PTFE-coated silicone cap liner seal
- Fully compatible with GL 25 or 45 threads and therefore highly versatile
- The matching PFA pouring ring permits drip-free pouring and replacement PTFE faced silicone cap liners (platinum-cured silicone) are available separately
Specifications
Specifications
| Description | Screw cap |
| For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Reservoir cap for supplying laboratory instruments; Port assemblies for spinner flasks, and cell culture vessels; Single port vent for autoclaving media in large bottles; Transport and shipping |
| Material | PFA, PTFE, Silicone |
| Quantity | 5/Pk. |
| Thread Size (Metric) | GL 45 |
| Diameter (Metric) | 51 mm |
| Autoclavable | Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.