Learn More
DWK Life Sciences DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
Brand:  DWK Life Sciences 292040209
9.85 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15202119
  • £12.15 / Pack of 10
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
  • Made from polyethylene
  • Octagonal shaped head
Specifications
Specifications
Storage of reagents, and chemicals
10/Pk.
No
ST 7/16
Polyethylene