missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
£3.00 - £23.30
Specifications
Autoclavable | No |
---|---|
Material | Polyethylene |
For Use With (Application) | Storage of reagents, and chemicals |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10603992
|
DWK Life Sciences
292041205 |
ST 45/40 |
£3.70
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
10613992
|
DWK Life Sciences
292041308 |
ST 60/46 |
£4.85
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15262119
|
DWK Life Sciences
292040903 |
ST 29/32 |
£23.30
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15242119
|
DWK Life Sciences
292040706 |
ST 19/26 |
£19.25
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15232119
|
DWK Life Sciences
292040603 |
ST 14/23 |
£14.40
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10161501
|
DWK Life Sciences
292041102 |
ST 34/45 |
£3.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10385531
|
DWK Life Sciences
292041608 |
ST 85/55 |
£6.95
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15212119
|
DWK Life Sciences
292040303 |
ST 10/19 |
£17.95
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15222119
|
DWK Life Sciences
292040406 |
ST 12/21 |
£21.70
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15252119
|
DWK Life Sciences
292040809 |
ST 24/29 |
£18.55
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15202119
|
DWK Life Sciences
292040209 |
ST 7/16 |
£12.15
Pack of 10 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
- Made from polyethylene
- Octagonal shaped head
Specifications
No | |
Polyethylene | |
Storage of reagents, and chemicals |