Learn More
DWK Life Sciences DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
Brand:  DWK Life Sciences 292041308
Code : NEW
Additional Details : Weight : 0.03800kg
 View more versions of this product
Product Code. 10613992
  • £4.85 / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
  • Made from polyethylene
  • Octagonal shaped head
Specifications
Specifications
Storage of reagents, and chemicals
1 Ea.
No
ST 60/46
Polyethylene