Learn More
DWK Life Sciences DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
Brand:  DWK Life Sciences 292041608
Code : NEW
Additional Details : Weight : 0.06400kg
 View more versions of this product
Product Code. 10385531
  • £6.95 / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
  • Made from polyethylene
  • Octagonal shaped head
Specifications
Specifications
Storage of reagents, and chemicals
1 Ea.
No
ST 85/55
Polyethylene