missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Pear Shape Flask, with standard ground joint

Product Code. 15631265 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15631265 25 mL Pack of 10
15621545 10 mL Pack of 10
15251899 50 mL Pack of 10
15261899 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15631265 Supplier DWK Life Sciences Supplier No. 241951402
Add to basket
Add to basket
This item is not returnable. View return policy

Pear shape flasks are used for processing small quantities of substances.

Due to their shape,pear shape flasks are particularly suitable for processing very small quantities. The standard ground joint allows a precise connection to other ground joint components such as columns, thermometers, drip funnels, etc.

  • Conical shape makes it ideal for small-scale reactions
  • Tight closure due to standard ground joint
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Base 40 mm
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Small-scale reactions; Distillation; Extraction; Connection to other glass vessels; Heating of small amounts; Suitable for heating baths and mantles; Evaporation of liquids; Suitable for mounting on tripods
No. per Pack 10/Pk.
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.