missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Organ Storage Jar, without stopper
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 10
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Organ glasses have a wide neck opening through which organs can be inserted for intermediate storage. A cover and thus the permanent storage is not foreseen in these vessels.
- Without lid
- Fired-on, highly durable white ceramic
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| For Use With (Application) | Storage of organs; Supplied without stopper |
| Material | Borosilicate Glass (3.3) |
| Quantity | 10/Pk. |
| Diameter (Metric) | 54 mm |
| Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction