missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT

Product Code. 11757008 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
11757008 GL14 Pack of 10
10434301 GL 45 Pack of 10
11767198 GL 18 Pack of 10
11737118 GL 25 Pack of 10
11787118 GL 32 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11757008 Supplier DWK Life Sciences Supplier No. 292270508
In Stock
Add to basket
Edge
Add to basket

PBT screw cap with an open top and high temperature and chemical resistance.

Open topped cap with central aperture suitable for use with bromobutyl stoppers, silicone piercing septa or glassware connections. More chemically resistant than PP cap.
  • Compatible with GL bottle threads
  • Red PBT offers high temperature and chemical resistance
  • Glass reinforced PBT offers high strength
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Diameter (Metric) Inner Bore Size 9.5 mm
For Use With (Application) Aseptic reagent delivery and culture sampling using a syringe and needle
Material PBT
Quantity 10/Pk.
Screw Material Polybutylene Terephthalate (PBT) with glass fiber
Thread Size (Metric) GL14
Closure Material PBT with Glass Fiber
Diameter (Metric) 20 mm
Autoclavable Autoclavable
Product Type Open Top Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.