missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT

Product Code. 11737118 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
11737118 GL 25 Pack of 10
10434301 GL 45 Pack of 10
11767198 GL 18 Pack of 10
11757008 GL14 Pack of 10
11787118 GL 32 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11737118 Supplier DWK Life Sciences Supplier No. 292270902
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT features an open top with central aperture for use with bromobutyl stoppers, silicone piercing septa or glassware connections. Made from glass-reinforced red PBT for excellent temperature and chemical resistance.

  • Open topped cap with central aperture compatible with bromobutyl stoppers, silicone piercing septa and glassware connections
  • Red PBT material offers excellent temperature and chemical resistance, superior to standard PP caps
  • Glass-reinforced PBT provides high mechanical strength
  • Compatible with standard GL bottle threads
  • Autoclavable for repeated sterilisation use

Specifications

Color Red
Diameter (Metric) Inner Bore Size 15 mm
For Use With (Application) Aseptic reagent delivery and culture sampling using a syringe and needle
Material PBT
Quantity 10/Pk.
Screw Material Polybutylene Terephthalate (PBT) with glass fiber
Sterility Non-sterile
Thread Size (Metric) GL 25
Closure Material PBT with Glass Fiber
Diameter (Metric) 33 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Open Top Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.