Learn More
DWK Life Sciences DURAN™ Multi-Connector Cap, GL 45, Tubing Connectors, for peristaltic tubing, 2, 3, 4, or 5 tubing connectors, Red PBT screw cap
Product Code. 16474460 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Connector Cap (2-port)
Connector Cap (3-port)
Connector Cap (4-port)
Connector Cap (5-port)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
16474460 Connector Cap (4-port) Each
16454954 Connector Cap (2-port) Each
16464460 Connector Cap (3-port) Each
16464954 Connector Cap (5-port) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16474460 Supplier DWK Life Sciences Supplier No. 292652808
Add to basket
Add to basket
2, 3, 4, or 5 peristaltic tubing connector caps for GL 45 DURAN™ bottles.
The reusable DURAN™ GL 45 Connection Cap System is ideal for aseptic fluid transfer into or out of DURAN™ bottles using peristaltic pump tubing. The screw caps will fit all DURAN™ Original GL 45 laboratory bottles 100 mL and larger. The barbed tubing connectors (4.2 mm outer diameter) accept narrow BPT peristaltic pump tubing with a 3.2 mm (1/8) inner diameter.
  • Caps have a GL 45 thread that fit DURAN™ bottles
  • Barbed male hose connector are suitable for attaching peristaltic PBT tubing with a 4.2 mm inner diameter
  • Available with 2, 3, 4, or 5 stainless steel (316L) barbed tubing connectors
  • Autoclavable at 121° C or 134°C
  • Construction materials are certified as approved for food contact
  • Reusable with a long service life
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Media supply for small bioreactors; Fed-batch bioreactor; Perfusion circuit for cell culture; Oxygenation/aeration of very small samples; Use in a chemistry micro-reactor
Product Type Connector Cap (4-port)