missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Microbiology Bottle Unground Wide Neck
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
Description
The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a wide conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.
- Wide neck for rubber stoppers
- Wide neck for easier filling and cleaning
- Retrace code
- Fired-on, highly durable white print
- Autoclavable at 121° C or 134°C
- Supplied without stopper
Specifications
Diameter (Metric) | 290 mm |
Capacity (Metric) | 20,000 mL |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel |
Material | Borosilicate Glass (3.3) |
Quantity | 1 Ea. |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'