Läs mer
DWK Life Sciences DURAN™ Laboratory Bottle, Square, Clear, with DIN 168-1 Thread, Graduated, without Screw Cap or Pouring Ring
Handla allt< titel> ProdukterBeskrivning
DURAN™ Square laboratory bottles have a space saving shape that offers the benefit of close and efficient packing. With up to a 20 % saving of space compared to traditional round bottles. Ideal for the shipping and storage of high-demand materials. For reagent supply to instruments where bench space is at a premium. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ square bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Ergonomic handling, high stability and good stackability due to angular shape
- Saves space and costs in autoclaves, drying chambers, ovens, storage or washers
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifikationer
Specifikationer
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Liquid handling reservoir for instruments; Transport and shipping; Sample collection; Storage of media and reagents; Archival storage of samples; Preparation of media and reagents; Culturing vessel |
| Includes | Without screw cap and pouring ring |
| Material | Borosilicate Glass (3.3) |
| Quantity | 10/Pk. |
| Thread Size (Metric) | GL 45 |
| Diameter (Metric) | 78 mm |
| Autoclavable | Autoclavable |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.