Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, flat, diamond-ground
Shop All DWK Life Sciences ProductsDescription
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
- Flat
- Diamond-ground
- hollow glass stopper
Specifications
Specifications
| For Use With (Application) | Closure of vessels with standard ground joints |
| Quantity | 100/Pk. |
| Size | NS 14/23 |
| Autoclavable | Autoclavable |
| Material | Borosilicate Glass |