Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, flat, diamond-ground
Product Code. 15814558 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15814558 NS 24/29 Pack of 100
10079940 NS 10/19 Pack of 100
10460873 NS 12/21 Pack of 100
15991521 NS 19/26 Pack of 100
15901531 NS 29/32 Pack of 100
10768631 NS 34/35 Each
15931531 NS 45/40 Pack of 10
15971521 NS 14/23 Pack of 100
15931521 NS 7/16 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15814558 Leverandør DWK Life Sciences Leverandørnr. 286127802
  • ONLINE EXCLUSIVE PRICE
    £270.38
    £256.63 / Pack of 100
    Save £13.75
    5% Off
Estimated Shipment: 02-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
Hollow glass stopper for sealing.
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Flat
  • Diamond-ground
  • hollow glass stopper
Tekniske data
For Use With (Application) Closure of vessels with standard ground joints
Quantity 100/Pk.
Size NS 24/29
Autoclavable Autoclavable
Material Borosilicate Glass
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.