Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, GLS 80™ Thread, Graduated, Bottle Only
Shop All DWK Life Sciences ProductsDescription
DURAN™ wide mouth GLS 80™ laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ wide mouth GLS 80™ are supplied complete with blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Specifications
Specifications
| Capacity (Metric) | 5000 mL |
| For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
| Includes | Without screw cap and pouring ring from PP (blue) |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Thread Size (Metric) | GLS 80 |
| Closure Material | Polypropylene |
| Diameter (Metric) | 182 mm |
| Autoclavable | Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.