missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ DURAN™ GLS 80™ Laboratory Bottle, Double Walled (Jacketed), Wide Mouth, Clear, Graduated

Product Code. 15476834 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15476834 1000 mL Each
15443253 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476834 Supplier DWK LIFE SCIENCES™ Supplier No. 242565456
  • ONLINE EXCLUSIVE PRICE
    £854.00
    £600.81 / Pack of 1
    Save £253.19
    29% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The GLS 80™ Laboratory Bottle, Double Walled (Jacketed), Wide Mouth, Clear, Graduated features an integral jacket that isolates contents from the external environment. Circulate heated or cooled liquids through the jacket for precise temperature control.

  • Integral jacket isolates bottle contents from the external environment for precise temperature control
  • Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw-topped bottle
  • Wide GLS 80 mouth provides easy access to bottle contents
  • 10 mm PTFE inlet/outlet hose connectors positioned around the jacket ensure consistent heating or cooling
  • Easy-to-read graduated volume scale on the inner vessel
  • Removable GL 18 outlet hose connectors for increased safety
  • Sealable and more flexible alternative to open-topped jacketed beakers

Specifications

Capacity (Metric) 1000 mL
Color Clear
For Use With (Application) Simple, low cost, jacketed chemostat or bioreactor for temperature-controlled reactions such as enzyme digestions; Storage at a constant temperature; Working with endo- or exo-thermic reagents; Large cold trap or condenser when used with dry ice in the inner container
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) GLS 80
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Material Polypropylene
Diameter (Metric) 110 mm
Autoclavable Autoclavable
Capacity (English) 32 oz.
Shape Round
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.