Learn More
Lenz Laborglasinstrumente™ Expansion Adapter with Ground Joint
Provides for secure and easy connection from a small cone to a large socket in glass tubing. Lenz Laborglasinstrumente™ Expansion Adapters with Ground Joints are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01504529
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 10323331
  • £22.00 / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12257
  • High taper accuracy of the cones
  • Minimized need for grease
  • Excellent tightness
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
DIN 12257 and in addition to DIN standard
Duran Glass
1 Ea.
NS 29/32
Expansion Adapter
NS 45/40